Gene Transcripts
- Gene Symbol
- M1AP
- Entrez Gene ID
- 130951
- Gene Name
- meiosis 1 associated protein
- Gene Aliases
- C2orf65, D6Mm5e, SPATA37
- Chromosome Location
- Chr. 2: 74557640 - 74648338 on Build GRCh38
- UniGene ID
- Hs.348645
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281295.1 |
NP_001268224.1
|
4, 5 |
721 |
|
NM_001281296.1 |
NP_001268225.1
|
4, 5 |
721 |
|
NM_001321739.1 |
NP_001308668.1
|
4, 5 |
721 |
|
NM_138804.4 |
NP_620159.2
|
4, 5 |
712 |
|
XM_006711946.3 |
XP_006712009.1
|
4, 5 |
1057 |
|
XM_011532548.2 |
XP_011530850.1
|
4, 5 |
1058 |
|
XM_011532549.2 |
XP_011530851.1
|
4, 5 |
1059 |
|
XM_011532550.2 |
XP_011530852.1
|
4, 5 |
668 |
|
XM_011532551.2 |
XP_011530853.1
|
4, 5 |
667 |
|
XM_011532553.2 |
XP_011530855.1
|
4, 5 |
1059 |
|
GenBank mRNA |
AK300188.1 |
- |
4, 5 |
713 |
|
BC014602.2 |
- |
4, 5 |
690 |
4821635 |
BC025997.1 |
- |
4, 5 |
715 |
4823281 |
View Full Product Details