Gene Transcripts

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
Chromosome Location
Chr. 3: 97821930 - 97944986 on Build GRCh38
UniGene ID
Hs.733378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153605.3 NP_705833.3 4 6739
XM_005247117.4 XP_005247174.1 3 6442
GenBank mRNA BX648353.1 - 1 789

View Full Product Details