Gene Transcripts
- Gene Symbol
- LINC00526
- Entrez Gene ID
- 147525
- Gene Name
- long intergenic non-protein coding RNA 526
- Gene Aliases
- C18orf18, HsT959
- Chromosome Location
- Chr. 18: 5236724 - 5238029 on Build GRCh38
- UniGene ID
- Hs.657197
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026849.1 |
|
1 |
639 |
|
GenBank mRNA |
AK292743.1 |
- |
1 |
1216 |
|
BC010538.1 |
- |
1 |
639 |
|
View Full Product Details