Gene Transcripts

Gene Symbol
NIPAL1
Entrez Gene ID
152519
Gene Name
NIPA like domain containing 1
Gene Aliases
NPAL1
Chromosome Location
Chr. 4: 48016772 - 48040173 on Build GRCh38
UniGene ID
Hs.134190
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207330.2 NP_997213.1 4, 5 525
XM_017007784.1 XP_016863273.1 4, 5 482
GenBank mRNA AK095299.1 - 4, 5 484
BC067881.1 - 4, 5 523 30345943
CB960388.1 - 4, 5 518
CR749484.1 - 4, 5 524

View Full Product Details