Gene Transcripts

Gene Symbol
LCN6
Entrez Gene ID
158062
Gene Name
lipocalin 6
Gene Aliases
LCN5, UNQ643, hLcn5
Chromosome Location
Chr. 9: 136744017 - 136748528 on Build GRCh38
UniGene ID
Hs.522504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198946.2 NP_945184.1 4 387
GenBank mRNA AF303084.1 - 4 384
AY301269.1 - 4 342
AY358598.1 - 4 367
BC062746.1 - 4 388 6452008

View Full Product Details