Gene Transcripts

Gene Symbol
RBM46
Entrez Gene ID
166863
Gene Name
RNA binding motif protein 46
Gene Aliases
CT68
Chromosome Location
Chr. 4: 154781250 - 154828813 on Build GRCh38
UniGene ID
Hs.133095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277171.1 NP_001264100.1 4 1562
NM_001277173.1 NP_001264102.1 4 1565
NM_144979.4 NP_659416.1 4 1562
XM_006714119.2 XP_006714182.1 4 1559
XM_006714120.1 XP_006714183.1 4 1559
XM_011531692.1 XP_011529994.1 4 1513
XM_011531693.1 XP_011529995.1 4 1565
XM_011531694.1 XP_011529996.1 4 1416
XM_011531695.2 XP_011529997.1 4 1461
XM_011531696.1 XP_011529998.1 4 1416
XM_011531697.2 XP_011529999.1 4 1559
GenBank mRNA AK125877.1 - 4 1565
AK302720.1 - 4 1499
BC028588.2 - 4 1486 4830259

View Full Product Details