Gene Transcripts

Gene Symbol
AES
Entrez Gene ID
166
Gene Name
amino-terminal enhancer of split
Gene Aliases
AES-1, AES-2, ESP1, GRG, GRG5, Grg-5, TLE5
Chromosome Location
Chr. 19: 3052910 - 3063107 on Build GRCh38
UniGene ID
Hs.515053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130.5 NP_001121.2 6 524
NM_198969.1 NP_945320.1 6 591
NM_198970.1 NP_945321.1 6 524
XM_006722664.1 XP_006722727.1 6 732
GenBank mRNA AF072902.1 - 6 427
AF269289.1 - 1 40
AK094591.1 - 6 733
AK095154.1 - 2 2045
AK314713.1 - 6 430
BC113735.1 - 6 578 8322787
BC113737.1 - 6 578 8322789
BG696926.1 - 2 108
BG913049.1 - 6 511
BM550097.1 - 481
BM908356.1 - 552
U04241.1 - 6 391
X73357.1 - 6 345
X73358.1 - 5 315

View Full Product Details