Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000112.3 | NP_000103.2 | 2 | 349 | |
XM_017009191.1 | XP_016864680.1 | 2 | 357 | ||
GenBank mRNA | AK290358.1 | - | 2 | 172 | |
AK312596.1 | - | 2 | 300 | ||
BC059390.1 | - | 2 | 203 | 30341764 | |
U14528.1 | - | 1 | 108 |