Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001398.2 | NP_001389.2 | 7 | 689 | |
XM_017026448.1 | XP_016881937.1 | 7 | 639 | ||
GenBank mRNA | AK126566.1 | - | 4 | 3088 | |
AK291860.1 | - | 7 | 624 | ||
BC011792.2 | - | 7 | 606 | 4300082 | |
BC014786.1 | - | 4 | 607 | ||
BC017408.1 | - | 7 | 630 | 4306900 | |
U16660.1 | - | 7 | 631 |