Gene Transcripts
- Gene Symbol
- MYRFL
- Entrez Gene ID
- 196446
- Gene Name
- myelin regulatory factor-like
- Gene Aliases
- C12orf15, C12orf28, bcm1377
- Chromosome Location
- Chr. 12: 69824518 - 69958726 on Build GRCh38
- UniGene ID
- Hs.253773
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_182530.2 |
NP_872336.2
|
14, 15 |
1955 |
|
XM_011539052.2 |
XP_011537354.1
|
13, 14 |
2436 |
|
XM_011539055.1 |
XP_011537357.1
|
12, 13 |
1423 |
|
XM_017018967.1 |
XP_016874456.1
|
13, 14 |
2237 |
|
XM_017018968.1 |
XP_016874457.1
|
14, 15 |
2691 |
|
XM_017018969.1 |
XP_016874458.1
|
13, 14 |
2171 |
|
XM_017018970.1 |
XP_016874459.1
|
12, 13 |
1981 |
|
XM_017018971.1 |
XP_016874460.1
|
14, 15 |
2760 |
|
XM_017018972.1 |
XP_016874461.1
|
14, 15 |
2759 |
|
XM_017018973.1 |
XP_016874462.1
|
10, 11 |
1652 |
|
GenBank mRNA |
AK057785.1 |
- |
1, 2 |
72 |
|
View Full Product Details