Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130047.1 | NP_001123519.1 | 7, 8 | 1042 | |
NM_152715.3 | NP_689928.3 | 7, 8 | 1169 | ||
XM_017017326.1 | XP_016872815.1 | 8, 9 | 1268 | ||
GenBank mRNA | AK295673.1 | - | 7, 8 | 1160 | |
AK313324.1 | - | 7, 8 | 1064 | ||
AY398644.1 | - | 7, 8 | 1152 | ||
BC020501.1 | - | 7, 8 | 1078 | ||
BC120988.1 | - | 6, 7 | 992 | 40116354 | |
BC120989.1 | - | 6, 7 | 992 | 40116355 |