Gene Transcripts
- Gene Symbol
- LRWD1
- Entrez Gene ID
- 222229
- Gene Name
- leucine rich repeats and WD repeat domain containing 1
- Gene Aliases
- CENP-33, ORCA
- Chromosome Location
- Chr. 7: 102464883 - 102473168 on Build GRCh38
- UniGene ID
- Hs.274135
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317721.1 |
NP_001304650.1
|
9, 10 |
1441 |
|
NM_152892.2 |
NP_690852.1
|
9, 10 |
1423 |
|
XM_017011845.1 |
XP_016867334.1
|
9, 10 |
1378 |
|
GenBank mRNA |
AB209379.1 |
- |
5, 6 |
1990 |
|
AK290967.1 |
- |
9, 10 |
1322 |
|
AK313771.1 |
- |
9, 10 |
1306 |
|
AL133057.1 |
- |
9, 10 |
1373 |
|
BC009436.1 |
- |
8, 9 |
1114 |
|
BC018769.1 |
- |
8, 9 |
953 |
|
BC030547.1 |
- |
9, 10 |
1363 |
5170588 |
View Full Product Details