Gene Transcripts

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
Chromosome Location
Chr. 10: 274190 - 689710 on Build GRCh38
UniGene ID
Hs.432397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014974.2 NP_055789.1 11 1373
XM_005252426.3 XP_005252483.1 12 1532
XM_005252427.3 XP_005252484.1 11 1413
XM_005252428.4 XP_005252485.1 12 1583
XM_011519428.2 XP_011517730.1 12 1514
XM_011519429.2 XP_011517731.1 12 1512
XM_011519430.2 XP_011517732.1 11 1389
XM_011519431.2 XP_011517733.1 11 1389
XM_011519432.1 XP_011517734.1 11 1814
GenBank mRNA AB023151.2 - 11 1371
AK126063.1 - 12 1419
AK310613.1 - 8 1008
BC063313.1 - 11 1373 30349267

View Full Product Details