Gene Transcripts

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
Chromosome Location
Chr. 9: 116425225 - 117415039 on Build GRCh38
UniGene ID
Hs.601562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184734.1 NP_001171663.1 3 543
NM_001184735.1 NP_001171664.1 3 543
NM_014010.4 NP_054729.3 17 3099
NM_198186.3 NP_937829.3 3 864
NM_198187.3 NP_937830.3 3 543
NM_198188.2 NP_937831.1 3 543
GenBank mRNA AB014534.1 - 17 3099
AF116574.1 - 15 2320
AK024064.1 - 3 514
AK127350.1 - 16 2320
AK315025.1 - 15 2163
BC018759.2 - 11 1548
BC029272.1 - 3 864
BC093835.1 - 3 377
BC101667.1 - 3 376
BC143289.1 - 3 377
BC143292.1 - 3 377
BC143293.1 - 3 377
BC146756.1 - 17 3099 9007192
BM552790.1 - 3 526
BX649176.1 - 17 2434

View Full Product Details