Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr. 22: 27978014 - 28679865 on Build GRCh38
UniGene ID
Hs.387856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145418.1 NP_001138890.1 4 836
XM_005261405.2 XP_005261462.1 4 836
XM_006724171.3 XP_006724234.1 3 9614
XM_011530018.2 XP_011528320.1 4 1015
XM_011530019.2 XP_011528321.1 4 836
XM_011530020.1 XP_011528322.1 4 836
XM_011530021.2 XP_011528323.1 4 836
XM_011530022.1 XP_011528324.1 4 836
XM_017028673.1 XP_016884162.1 4 809
GenBank mRNA AB665284.1 - 4 784

View Full Product Details