Gene Transcripts
- Gene Symbol
- ANP32C
- Entrez Gene ID
- 23520
- Gene Name
- acidic nuclear phosphoprotein 32 family member C
- Gene Aliases
- PP32R1
- Chromosome Location
- Chr. 4: 164197007 - 164197711 on Build GRCh38
- UniGene ID
- Hs.661161
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012403.1 |
NP_036535.1
|
1 |
406 |
|
View Full Product Details