Gene Transcripts

Gene Symbol
PADI4
Entrez Gene ID
23569
Gene Name
peptidyl arginine deiminase 4
Gene Aliases
PAD, PAD4, PADI5, PDI4, PDI5
Chromosome Location
Chr. 1: 17308195 - 17364004 on Build GRCh38
UniGene ID
Hs.522969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012387.2 NP_036519.2 11, 12 1345
XM_011541150.1 XP_011539452.1 9, 10 1157
XM_011541152.1 XP_011539454.1 8, 9 877
XM_011541157.1 XP_011539459.1 8, 9 824
GenBank mRNA AB017919.1 - 11, 12 1343
AK290507.1 - 11, 12 1343
AK314839.1 - 11, 12 1345
BC025718.1 - 11, 12 1329 5228777

View Full Product Details