Gene Transcripts

Gene Symbol
HCG27
Entrez Gene ID
253018
Gene Name
HLA complex group 27 (non-protein coding)
Gene Aliases
bCX101P6.9, bPG299F13.9, bQB115I13.2
Chromosome Location
Chr. 6: 31197760 - 31203968 on Build GRCh38
UniGene ID
Hs.659818
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026791.1 2 382
GenBank mRNA AK097442.1 - 2 381
BC041465.2 - 1 261
BC104432.1 - 2 303
BC104433.1 - 2 305
DB027671.1 - 2 381

View Full Product Details