Gene Transcripts

Gene Symbol
TBC1D28
Entrez Gene ID
254272
Gene Name
TBC1 domain family member 28
Gene Aliases
Chromosome Location
Chr. 17: 18634434 - 18644431 on Build GRCh38
UniGene ID
Hs.434123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039397.2 NP_001034486.2 10 981
GenBank mRNA AK097563.1 - 9 1269
BC112012.1 - 8 647 8327480
BC112016.1 - 8 647 8327484

View Full Product Details