Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 8 | 1036 | |
NM_015386.2 | NP_056201.2 | 8 | 1036 | ||
XM_011522981.2 | XP_011521283.1 | 9 | 1196 | ||
GenBank mRNA | AB088369.1 | - | 9 | 1172 | |
AK022874.1 | - | 8 | 1016 | ||
AK026435.1 | - | 4 | 330 | ||
AK096557.1 | - | 6 | 754 | ||
AK293235.1 | - | 5 | 580 | ||
AK297557.1 | - | 9 | 979 | ||
AL050101.1 | - | 4 | 338 | ||
AU125729.1 | - | 5 | 514 | ||
BC006306.2 | - | 5 | 552 | ||
BC013347.2 | - | 4 | 344 | ||
BC027726.2 | - | 8 | 1010 | ||
BC072438.1 | - | 8 | 1016 | 6011802 | |
BC128144.1 | - | 8 | 1013 | ||
BC128145.1 | - | 8 | 1013 | ||
BX648010.1 | - | 6 | 1704 |