Gene Transcripts

Gene Symbol
TEX40
Entrez Gene ID
25858
Gene Name
testis expressed 40
Gene Aliases
C11orf20
Chromosome Location
Chr. 11: 64300391 - 64304767 on Build GRCh38
UniGene ID
Hs.97174
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039496.1 NP_001034585.1 2, 3 360
GenBank mRNA AL117564.1 - 2, 3 363

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
Chromosome Location
Chr. 11: 64291722 - 64304769 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_133662.1 8, 9 2347

View Full Product Details