Gene Transcripts

Gene Symbol
WHRN
Entrez Gene ID
25861
Gene Name
whirlin
Gene Aliases
CIP98, DFNB31, PDZD7B, USH2D, WI
Chromosome Location
Chr. 9: 114402078 - 114505509 on Build GRCh38
UniGene ID
Hs.93836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083885.2 NP_001077354.2 12 2053
NM_001173425.1 NP_001166896.1 12 3214
NM_015404.3 NP_056219.3 12 3217
XM_011518484.1 XP_011516786.1 13 3303
XM_011518485.1 XP_011516787.1 14 3151
XM_011518486.1 XP_011516788.1 13 3300
XM_011518487.1 XP_011516789.1 12 3177
XM_011518488.1 XP_011516790.1 11 3060
XM_011518495.1 XP_011516797.1 10 1555
GenBank mRNA AB040959.1 - 12 2731
AK022854.1 - 8 1931
AL110228.1 - 12 2011
BC011918.2 - 3 326
BC014524.1 - 3714
BC136416.1 - 12 2963 9020404
BC142614.1 - 12 3217 40147287
BC142684.1 - 12 3192 40148465

View Full Product Details