Gene Transcripts

Gene Symbol
SUSD5
Entrez Gene ID
26032
Gene Name
sushi domain containing 5
Gene Aliases
Chromosome Location
Chr. 3: 33150041 - 33219215 on Build GRCh38
UniGene ID
Hs.196647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015551.1 NP_056366.1 5 1167
XM_005265034.3 XP_005265091.1 4 715
XM_017006137.1 XP_016861626.1 5 1117
GenBank mRNA AB011099.1 - 5 1167
AK295869.1 - 5 762

View Full Product Details