Gene Transcripts

Gene Symbol
EPC2
Entrez Gene ID
26122
Gene Name
enhancer of polycomb homolog 2
Gene Aliases
EPC-LIKE
Chromosome Location
Chr. 2: 148644780 - 148788491 on Build GRCh38
UniGene ID
Hs.23270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015630.3 NP_056445.3 7 1150
XM_011510941.2 XP_011509243.1 7 1361
XM_011510943.2 XP_011509245.1 6 1034
GenBank mRNA AF286904.1 - 7 1142
AK001433.1 - 5 741
AK125789.1 - 8 1219
BC093818.1 - 7 1062 7939663
BC093820.1 - 7 1062 7939665

View Full Product Details