Gene Transcripts

Gene Symbol
FAM50B
Entrez Gene ID
26240
Gene Name
family with sequence similarity 50 member B
Gene Aliases
D6S2654E, X5L
Chromosome Location
Chr. 6: 3831901 - 3851320 on Build GRCh38
UniGene ID
Hs.140944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012135.2 NP_036267.1 2 942
XM_017010729.1 XP_016866218.1 2 925
GenBank mRNA BC001261.1 - 2 920
Y18503.1 - 2 910

View Full Product Details