Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012434.4 | NP_036566.1 | 10 | 1419 | |
XM_005248710.3 | XP_005248767.1 | 10 | 1441 | ||
XM_005248711.2 | XP_005248768.1 | 10 | 1673 | ||
XM_011535750.2 | XP_011534052.1 | 9 | 1229 | ||
XM_017010730.1 | XP_016866219.1 | 9 | 1585 | ||
XM_017010731.1 | XP_016866220.1 | 9 | 1435 | ||
GenBank mRNA | AF244577.1 | - | 11 | 1676 | |
AJ387747.1 | - | 10 | 1561 | ||
AK025880.1 | - | 4 | 332 | ||
AK026921.1 | - | 2 | 84 | ||
AK075320.1 | - | 11 | 1475 | ||
AK315789.1 | - | 10 | 1377 | ||
BC020961.2 | - | 10 | 1378 | 3847279 |