Gene Transcripts
- Gene Symbol
- EIF2AK1
- Entrez Gene ID
- 27102
- Gene Name
- eukaryotic translation initiation factor 2 alpha kinase 1
- Gene Aliases
- HCR, HRI
- Chromosome Location
- Chr. 7: 6022247 - 6059229 on Build GRCh38
- UniGene ID
- Hs.520205
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134335.1 |
NP_001127807.1
|
4, 5 |
603 |
|
NM_014413.3 |
NP_055228.2
|
4, 5 |
603 |
|
GenBank mRNA |
AB037790.1 |
- |
4, 5 |
528 |
|
AF100784.1 |
- |
4, 5 |
492 |
|
AF147094.1 |
- |
4, 5 |
538 |
|
AF183414.1 |
- |
4, 5 |
490 |
|
AF255050.1 |
- |
4, 5 |
548 |
|
AK075192.1 |
- |
4, 5 |
531 |
|
AK290327.1 |
- |
4, 5 |
536 |
|
AK295712.1 |
- |
3, 4 |
404 |
|
AL136563.1 |
- |
4, 5 |
508 |
|
AL834494.1 |
- |
4, 5 |
508 |
|
BC006524.2 |
- |
4, 5 |
535 |
2989989 |
View Full Product Details