Gene Transcripts

Gene Symbol
STK36
Entrez Gene ID
27148
Gene Name
serine/threonine kinase 36
Gene Aliases
FU
Chromosome Location
Chr. 2: 218672026 - 218702717 on Build GRCh38
UniGene ID
Hs.471404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243313.1 NP_001230242.1 26 3432
NM_015690.4 NP_056505.2 26 3495
XM_005246464.1 XP_005246521.1 26 3383
XM_011510959.1 XP_011509261.1 26 3468
XM_011510960.1 XP_011509262.1 26 3436
XM_011510961.1 XP_011509263.1 17 2066
XM_017003804.1 XP_016859293.1 26 3320
XM_017003805.1 XP_016859294.1 25 3172
GenBank mRNA AB033104.1 - 26 3389
AF200815.1 - 25 3216
AK022692.1 - 5 655
AL133630.1 - 6 751
BC026158.2 - 26 3435 4839214

View Full Product Details