Gene Transcripts
- Gene Symbol
- NFU1
- Entrez Gene ID
- 27247
- Gene Name
- NFU1 iron-sulfur cluster scaffold
- Gene Aliases
- CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU
- Chromosome Location
- Chr. 2: 69396113 - 69438123 on Build GRCh38
- UniGene ID
- Hs.430439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002755.2 |
NP_001002755.1
|
7, 8 |
927 |
|
NM_001002756.2 |
NP_001002756.1
|
6, 7 |
881 |
|
NM_015700.3 |
NP_056515.2
|
7, 8 |
1017 |
|
NR_045631.1 |
|
6, 7 |
791 |
|
XM_017003808.1 |
XP_016859297.1
|
7, 8 |
730 |
|
GenBank mRNA |
AA625581.1 |
- |
1, 2 |
168 |
|
AF132967.1 |
- |
7, 8 |
729 |
|
AI129426.1 |
- |
1, 2 |
177 |
|
AJ132584.1 |
- |
7, 8 |
805 |
|
AK300700.1 |
- |
7, 8 |
736 |
|
AK314004.1 |
- |
7, 8 |
760 |
|
AY286306.1 |
- |
7, 8 |
774 |
|
AY286307.1 |
- |
7, 8 |
864 |
|
AY335194.1 |
- |
7, 8 |
920 |
|
BC113692.1 |
- |
7, 8 |
889 |
8322744 |
BC113694.1 |
- |
7, 8 |
889 |
8322746 |
BU178674.1 |
- |
6, 7 |
623 |
|
BU507991.1 |
- |
7, 8 |
732 |
|
View Full Product Details