Gene Transcripts
- Gene Symbol
- GPI
- Entrez Gene ID
- 2821
- Gene Name
- glucose-6-phosphate isomerase
- Gene Aliases
- AMF, GNPI, NLK, PGI, PHI, SA-36, SA36
- Chromosome Location
- Chr. 19: 34359592 - 34402413 on Build GRCh38
- UniGene ID
- Hs.466471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000175.3 |
NP_000166.2
|
1, 2 |
265 |
|
NM_001184722.1 |
NP_001171651.1
|
2, 3 |
412 |
|
NM_001289789.1 |
NP_001276718.1
|
2, 3 |
412 |
|
NM_001289790.1 |
NP_001276719.1
|
1, 2 |
265 |
|
XM_005258764.2 |
XP_005258821.1
|
2, 3 |
231 |
|
XM_006723148.2 |
XP_006723211.1
|
2, 3 |
182 |
|
XM_011526754.2 |
XP_011525056.1
|
3, 4 |
647 |
|
GenBank mRNA |
AK129884.1 |
- |
2, 3 |
196 |
|
AK293446.1 |
- |
1, 2 |
141 |
|
AK294396.1 |
- |
2, 3 |
360 |
|
AK301103.1 |
- |
1, 2 |
150 |
|
BC004982.1 |
- |
1, 2 |
133 |
2906270 |
DC299841.1 |
- |
1, 2 |
265 |
|
DC342043.1 |
- |
2, 3 |
412 |
|
K03515.1 |
- |
1, 2 |
140 |
|
X16539.1 |
- |
1, 2 |
128 |
|
View Full Product Details