Gene Transcripts

Gene Symbol
REC114
Entrez Gene ID
283677
Gene Name
REC114 meiotic recombination protein
Gene Aliases
C15orf60, CT147
Chromosome Location
Chr. 15: 73443158 - 73560015 on Build GRCh38
UniGene ID
Hs.743574
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042367.1 NP_001035826.1 5 586
XM_017022083.1 XP_016877572.1 4 378
GenBank mRNA AK131030.1 - 4 496
BC055412.1 - 5 567
BX113903.1 - 3 297

View Full Product Details