Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144877.2 | NP_001138349.1 | 12 | 1202 | |
NM_173690.4 | NP_775961.2 | 13 | 1271 | ||
GenBank mRNA | AK093983.1 | - | 12 | 1161 | |
BC104030.2 | - | 13 | 1199 | 40025626 | |
BC104031.1 | - | 12 | 1130 | 40025627 | |
BC104032.1 | - | 12 | 1130 | 40025629 |