Gene Transcripts

Gene Symbol
GPS1
Entrez Gene ID
2873
Gene Name
G protein pathway suppressor 1
Gene Aliases
COPS1, CSN1, SGN1
Chromosome Location
Chr. 17: 82051477 - 82057470 on Build GRCh38
UniGene ID
Hs.268530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321089.1 NP_001308018.1 10 1251
NM_001321090.1 NP_001308019.1 10 1239
NM_001321091.1 NP_001308020.1 10 1236
NM_001321092.1 NP_001308021.1 9 1035
NM_001321093.1 NP_001308022.1 10 1120
NM_004127.5 NP_004118.3 9 1047
NM_212492.2 NP_997657.1 9 1530
XM_005256350.1 XP_005256407.1 10 1219
XM_005256353.1 XP_005256410.1 10 1207
XM_005256359.1 XP_005256416.1 9 1011
XM_005256360.3 XP_005256417.1 9 1040
XM_011523569.1 XP_011521871.1 10 1072
XM_011523570.1 XP_011521872.1 10 1110
XM_017024524.1 XP_016880013.1 9 1026
XM_017024525.1 XP_016880014.1 10 1204
XM_017024526.1 XP_016880015.1 9 1026
XM_017024527.1 XP_016880016.1 9 1037
XM_017024528.1 XP_016880017.1 9 995
XM_017024529.1 XP_016880018.1 9 992
XM_017024530.1 XP_016880019.1 9 983
XM_017024531.1 XP_016880020.1 10 1057
XM_017024532.1 XP_016880021.1 9 1008
XM_017024533.1 XP_016880022.1 9 1024
XM_017024534.1 XP_016880023.1 10 1072
XM_017024535.1 XP_016880024.1 10 1057
GenBank mRNA AB209596.1 - 3 3167
AK026610.1 - 10 1082
AK093283.1 - 9 965
AK222508.1 - 9 1000
AK225248.1 - 9 997
AK289435.1 - 9 1150
AK297872.1 - 9 1023
BC000155.2 - 9 1003 2900944
BC064503.1 - 9 1442 6572943
BT009834.1 - 8 932
U20285.2 - 9 1027

View Full Product Details