Gene Transcripts
- Gene Symbol
- METTL5
- Entrez Gene ID
- 29081
- Gene Name
- methyltransferase like 5
- Gene Aliases
- HSPC133
- Chromosome Location
- Chr. 2: 169811757 - 169824931 on Build GRCh38
- UniGene ID
- Hs.470553
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293186.1 |
NP_001280115.1
|
2, 3 |
303 |
|
NM_001293187.1 |
NP_001280116.1
|
2, 3 |
217 |
|
NM_014168.3 |
NP_054887.2
|
1, 2 |
444 |
|
XM_011511023.1 |
XP_011509325.1
|
2, 3 |
244 |
|
GenBank mRNA |
AF161482.1 |
- |
2, 3 |
192 |
|
AF201938.1 |
- |
2, 3 |
261 |
|
AK001321.1 |
- |
2, 3 |
191 |
|
AL523293.3 |
- |
1, 2 |
426 |
|
BC000921.2 |
- |
1, 2 |
356 |
3445582 |
BC093014.1 |
- |
2, 3 |
199 |
4799279 |
CA337169.1 |
- |
2, 3 |
218 |
|
View Full Product Details