Gene Transcripts
- Gene Symbol
- HLA-DRA
- Entrez Gene ID
- 3122
- Gene Name
- major histocompatibility complex, class II, DR alpha
- Gene Aliases
- HLA-DRA1, MLRW
- Chromosome Location
- Chr. 6: 32439842 - 32445046 on Build GRCh38
- UniGene ID
- Hs.520048
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019111.4 |
NP_061984.2
|
3, 4 |
719 |
|
GenBank mRNA |
AK301134.1 |
- |
3, 4 |
616 |
|
AK313123.1 |
- |
3, 4 |
674 |
|
BC032350.1 |
- |
3, 4 |
650 |
5214156 |
BC071659.1 |
- |
3, 4 |
658 |
5162925 |
BG548634.1 |
- |
2, 3 |
345 |
|
CR457013.1 |
- |
3, 4 |
610 |
|
J00194.1 |
- |
3, 4 |
636 |
|
K01171.1 |
- |
3, 4 |
624 |
|
M60333.1 |
- |
3, 4 |
562 |
|
M60334.1 |
- |
3, 4 |
637 |
|
- Gene Symbol
- HLA-DQA1
- Entrez Gene ID
- 3117
- Gene Name
- major histocompatibility complex, class II, DQ alpha 1
- Gene Aliases
- CD, CELIAC1, DQ-A1, GSE, HLA-DQA
- Chromosome Location
- Chr. 6: 32637403 - 32654846 on Build GRCh38
- UniGene ID
- Hs.387679
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF522250.1 |
- |
|
630 |
|
View Full Product Details