Gene Transcripts

Gene Symbol
HNRNPC
Entrez Gene ID
3183
Gene Name
heterogeneous nuclear ribonucleoprotein C (C1/C2)
Gene Aliases
C1, C2, HNRNP, HNRPC, SNRPC
Chromosome Location
Chr. 14: 21209136 - 21269479 on Build GRCh38
UniGene ID
Hs.508848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077442.1 NP_001070910.1 2 380
NM_001077443.1 NP_001070911.1 2 380
NM_004500.3 NP_004491.2 3 406
NM_031314.2 NP_112604.2 3 406
XM_006720125.2 XP_006720188.1 3 1062
XM_011536708.1 XP_011535010.1 4 440
XM_011536709.2 XP_011535011.1 3 861
XM_011536710.2 XP_011535012.1 3 1062
XM_011536711.2 XP_011535013.1 3 1069
XM_011536712.1 XP_011535014.1 4 440
XM_017021252.1 XP_016876741.1 3 1069
XM_017021253.1 XP_016876742.1 3 860
GenBank mRNA AB209527.1 - 3 340
AK126950.1 - 2 3114
AK223517.1 - 3 345
AK225032.1 - 3 369
AK292619.1 - 5 683
AK297482.1 - 2 325
AK299923.1 - 3 351
AK302213.1 - 2 343
AK312338.1 - 3 351
AK312382.1 - 2 326
BC003394.1 - 3 337 3447724
BC007052.1 - 2 326 3686841
BC008364.1 - 3 344 3354131
BC008423.1 - 2 326 4102910
BC066932.1 - 2 325 4375450
BC089438.1 - 2 301 30555817
BC103758.1 - 2 325 6187512
BC108658.1 - 2 270 6501821
BX161480.1 - 2 275
BX247992.1 - 2 291
DB449514.1 - 3 407
M16342.1 - 3 284
M29063.1 - 3 341

View Full Product Details