Gene Transcripts
- Gene Symbol
- HNRNPL
- Entrez Gene ID
- 3191
- Gene Name
- heterogeneous nuclear ribonucleoprotein L
- Gene Aliases
- HNRPL, P/OKcl.14, hnRNP-L
- Chromosome Location
- Chr. 19: 38836378 - 38852339 on Build GRCh38
- UniGene ID
- Hs.644906
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005335.1 |
NP_001005335.1
|
4, 5 |
479 |
|
NM_001533.2 |
NP_001524.2
|
4, 5 |
713 |
|
XM_011526886.1 |
XP_011525188.1
|
4, 5 |
502 |
|
XM_011526887.1 |
XP_011525189.1
|
4, 5 |
546 |
|
XM_011526889.1 |
XP_011525191.1
|
4, 5 |
597 |
|
GenBank mRNA |
AB044547.1 |
- |
4, 5 |
708 |
|
AK292115.1 |
- |
4, 5 |
478 |
|
AK298384.1 |
- |
4, 5 |
549 |
|
AK301062.1 |
- |
4, 5 |
487 |
|
AK313652.1 |
- |
4, 5 |
648 |
|
BC069184.1 |
- |
4, 5 |
652 |
|
BI256815.1 |
- |
4, 5 |
479 |
|
X16135.1 |
- |
4, 5 |
637 |
|
View Full Product Details