Gene Transcripts

Gene Symbol
LRRIQ4
Entrez Gene ID
344657
Gene Name
leucine rich repeats and IQ motif containing 4
Gene Aliases
LRRC64
Chromosome Location
Chr. 3: 169812947 - 169838007 on Build GRCh38
UniGene ID
Hs.319484
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080460.1 NP_001073929.1 4 1389
XM_005247421.2 XP_005247478.1 5 1500
XM_006713613.3 XP_006713676.1 5 1748
XM_011512748.1 XP_011511050.1 5 1499
XM_011512749.1 XP_011511051.1 4 1326

View Full Product Details