Gene Transcripts

Gene Symbol
APOC3
Entrez Gene ID
345
Gene Name
apolipoprotein C3
Gene Aliases
APOCIII, HALP2
Chromosome Location
Chr. 11: 116829908 - 116833071 on Build GRCh38
UniGene ID
Hs.73849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000040.1 NP_000031.1 3 143
GenBank mRNA BC027977.1 - 3 141
M28613.1 - 2 127
X00567.1 - 2 97
X01388.1 - 3 168

View Full Product Details