Gene Transcripts
- Gene Symbol
- WDR86
- Entrez Gene ID
- 349136
- Gene Name
- WD repeat domain 86
- Gene Aliases
- Chromosome Location
- Chr. 7: 151377974 - 151410705 on Build GRCh38
- UniGene ID
- Hs.647083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284260.1 |
NP_001271189.1
|
2, 3 |
755 |
|
NM_001284261.1 |
NP_001271190.1
|
2, 3 |
256 |
|
NM_001284262.1 |
NP_001271191.1
|
2, 3 |
256 |
|
NM_198285.2 |
NP_938026.2
|
2, 3 |
755 |
|
XM_005249989.3 |
XP_005250046.1
|
2, 3 |
752 |
|
XM_005249990.4 |
XP_005250047.1
|
2, 3 |
752 |
|
XM_006715966.2 |
XP_006716029.1
|
2, 3 |
752 |
|
XM_011516144.2 |
XP_011514446.1
|
2, 3 |
752 |
|
XM_011516145.2 |
XP_011514447.1
|
2, 3 |
752 |
|
XM_011516146.2 |
XP_011514448.1
|
2, 3 |
752 |
|
XM_011516147.2 |
XP_011514449.1
|
2, 3 |
752 |
|
XM_011516148.1 |
XP_011514450.1
|
2, 3 |
240 |
|
XM_011516150.1 |
XP_011514452.1
|
3, 4 |
444 |
|
XM_011516151.2 |
XP_011514453.1
|
2, 3 |
752 |
|
XM_011516152.1 |
XP_011514454.1
|
2, 3 |
752 |
|
GenBank mRNA |
AK125347.1 |
- |
2, 3 |
205 |
|
AK296049.1 |
- |
2, 3 |
348 |
|
BC107125.2 |
- |
1, 2 |
152 |
40023951 |
BI757623.1 |
- |
2, 3 |
737 |
|
BX338490.2 |
- |
2, 3 |
678 |
|
View Full Product Details