Gene Transcripts

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
Chromosome Location
Chr. 10: 116324097 - 116380029 on Build GRCh38
UniGene ID
Hs.233407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198515.2 NP_940917.1 9 1014
XM_006717820.3 XP_006717883.1 9 1309
XM_017016206.1 XP_016871695.1 9 932
GenBank mRNA AJ457798.1 - 8 996
BC044830.1 - 9 1014 5166167

View Full Product Details