Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
Chromosome Location
Chr. 5: 66596348 - 67169595 on Build GRCh38
UniGene ID
Hs.595458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164664.1 NP_001158136.1 28 4125
NM_001290226.1 NP_001277155.1 26 3577
NM_001290227.1 NP_001277156.1 25 3148
NM_001297651.1 NP_001284580.1 25 3609
NM_015183.2 NP_055998.1 27 3318
XM_006714606.3 XP_006714669.1 26 3571
XM_006714610.2 XP_006714673.1 24 3357
XM_011543382.2 XP_011541684.1 30 4217
XM_011543384.2 XP_011541686.1 25 5276
XM_011543385.2 XP_011541687.1 25 6396
XM_011543386.2 XP_011541688.1 25 3227
XM_017009447.1 XP_016864936.1 31 4298
XM_017009448.1 XP_016864937.1 29 4181
XM_017009449.1 XP_016864938.1 26 3616
XM_017009450.1 XP_016864939.1 26 3310
XM_017009451.1 XP_016864940.1 25 3109
XM_017009452.1 XP_016864941.1 26 6625
XM_017009453.1 XP_016864942.1 25 13440
GenBank mRNA AB002301.1 - 17 2361
AY830839.1 - 27 3319
BX648796.1 - 2 156
BX648980.1 - 7 927

View Full Product Details