Gene Transcripts

Gene Symbol
KIFC3
Entrez Gene ID
3801
Gene Name
kinesin family member C3
Gene Aliases
Chromosome Location
Chr. 16: 57758217 - 57863053 on Build GRCh38
UniGene ID
Hs.23131
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130099.1 NP_001123571.1 13 1977
NM_001130100.1 NP_001123572.1 14 2033
NM_001318710.1 NP_001305639.1 14 1914
NM_001318711.1 NP_001305640.1 14 2075
NM_001318712.1 NP_001305641.1 12 1650
NM_001318713.1 NP_001305642.1 12 2043
NM_001318714.1 NP_001305643.1 15 2422
NM_001318715.1 NP_001305644.1 14 2011
NM_005550.3 NP_005541.3 14 2033
NR_134678.1 14 2217
XM_005255937.1 XP_005255994.1 16 2542
XM_005255944.1 XP_005256001.1 14 1896
XM_006721188.1 XP_006721251.1 17 2620
XM_011523075.1 XP_011521377.1 14 2200
XM_011523076.1 XP_011521378.1 14 2200
XM_011523077.1 XP_011521379.1 14 1999
XM_011523078.1 XP_011521380.1 14 1802
XM_011523079.1 XP_011521381.1 14 2137
XM_017023219.1 XP_016878708.1 17 2589
XM_017023220.1 XP_016878709.1 17 2766
XM_017023221.1 XP_016878710.1 16 2542
XM_017023222.1 XP_016878711.1 18 2784
XM_017023223.1 XP_016878712.1 14 1917
XM_017023224.1 XP_016878713.1 16 2451
XM_017023225.1 XP_016878714.1 12 3021
XM_017023226.1 XP_016878715.1 15 2294
XM_017023227.1 XP_016878716.1 15 2294
GenBank mRNA AB209290.1 - 14 1968
AF004426.1 - 10 1419
AK291737.1 - 13 1872
AK295926.1 - 9 995
AK296995.1 - 14 1913
AK299431.1 - 14 1975
AK302743.1 - 12 1650
AK303021.1 - 12 1971
AK304132.1 - 14 2021
AK307846.1 - 14 1991
BC001211.1 - 14 2002 3503847
BC008014.1 - 8 998
BC034234.1 - 15 2362 5296397
BC041132.1 - 14 1863 5760988
BC047051.1 - 14 2161 5297265

View Full Product Details