Gene Transcripts

Gene Symbol
SUMO4
Entrez Gene ID
387082
Gene Name
small ubiquitin-like modifier 4
Gene Aliases
IDDM5, SMT3H4, SUMO-4, dJ281H8.4
Chromosome Location
Chr. 6: 149400359 - 149401046 on Build GRCh38
UniGene ID
Hs.657168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002255.1 NP_001002255.1 1 247
GenBank mRNA AY340238.1 - 1 247
BC130305.1 - 1 241 40146336

View Full Product Details