Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289973.1 | NP_001276902.1 | 5 | 874 | |
NM_207411.4 | NP_997294.3 | 4 | 736 | ||
XM_017013401.1 | XP_016868890.1 | 3 | 1155 | ||
GenBank mRNA | AY358489.1 | - | 4 | 607 | |
AY534242.1 | - | 4 | 736 | ||
AY534243.1 | - | 5 | 728 |