Gene Transcripts

Gene Symbol
OR51B6
Entrez Gene ID
390058
Gene Name
olfactory receptor family 51 subfamily B member 6
Gene Aliases
HOR5'Beta6
Chromosome Location
Chr. 11: 5351508 - 5352446 on Build GRCh38
UniGene ID
Hs.553730
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004750.1 NP_001004750.1 1 762

View Full Product Details