Gene Transcripts

Gene Symbol
OR4F6
Entrez Gene ID
390648
Gene Name
olfactory receptor family 4 subfamily F member 6
Gene Aliases
OR15-15, OR4F12
Chromosome Location
Chr. 15: 101805720 - 101806658 on Build GRCh38
UniGene ID
Hs.553399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005326.1 NP_001005326.1 1 150
GenBank mRNA BC137002.1 - 1 171
BC137003.1 - 1 171 9020996

View Full Product Details