Gene Transcripts

Gene Symbol
PTX4
Entrez Gene ID
390667
Gene Name
pentraxin 4
Gene Aliases
C16orf38
Chromosome Location
Chr. 16: 1485939 - 1488467 on Build GRCh38
UniGene ID
Hs.553770
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013658.1 NP_001013680.1 3 1334

View Full Product Details