Gene Transcripts
- Gene Symbol
- IGFL2
- Entrez Gene ID
- 147920
- Gene Name
- IGF like family member 2
- Gene Aliases
- UNQ645, VPRI645
- Chromosome Location
- Chr. 19: 46078513 - 46195097 on Build GRCh38
- UniGene ID
- Hs.99376
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125754.1 |
- |
1 |
630 |
|
View Full Product Details