Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207581.3 | NP_997464.2 | 1 | 359 | |
XM_017022180.1 | XP_016877669.1 | 1 | 363 | ||
GenBank mRNA | AK303955.1 | - | 1 | 358 | |
BC137465.1 | - | 1 | 280 | 9021465 | |
BX537581.1 | - | 1 | 210 | ||
DB243076.1 | - | 1 | 358 | ||
DC393748.1 | - | 1 | 359 | ||
DQ489734.1 | - | 1 | 210 |